Key Documents
PL800GB-8
Plas-Labs butyl glove box gloves
for 1 piece glove set, size 8 (for 8 in. ports)
Select a Size
1 EACH
€716.00
€716.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
€716.00
About This Item
UNSPSC Code:
42132201
NACRES:
NB.35
€716.00
Please contact Customer Service for Availability
size
8 (for 8 in. ports)
General description
Plas-Labs black butyl rubber gloves for highest impermeability to water vapor, gases, and toxic fluids
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
Choose from one of the most recent versions:
Certificates of Analysis (COA)