Documents
PL800GA-BUT-7
Plas-Labs replacement butyl gloves
for 2 piece glove set, size 7 (for 8 in. ports)
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
material
butyl rubber
size
7 (for 8 in. ports)
feature
reusable
for use with glove box sleeves
packaging
pack of 1 pair
manufacturer/tradename
Plas-Labs 800GA-BUT-7
General description
Plas-Labs black butyl rubber gloves for highest impermeability to water vapor, gases, and toxic fluids
Note: Order sleeves, gloves, and glove over sleeve connectors separately.