Skip to Content
Merck
All Photos(2)

Documents

PL800GH

Plas-Labs Hypalon glove box gloves

size 9 (for 8 in. ports)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
42132201
NACRES:
NB.35

size

9 (for 8 in. ports)

description

32" long, 9.5" handsize. Sold as a pair.

Looking for similar products? Visit Product Comparison Guide

General description

Plas-Labs white Hypalon® rubber for resistance to abrasion, oxidizing chemicals, ozone and UV light

Note: Order sleeves, gloves, and glove over sleeve connectors separately.

Legal Information

HaackerCalc is a trademark of ChemSW, Inc.
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service