Skip to Content
Merck
All Photos(2)

Key Documents

769053

Sigma-Aldrich

Oxygen

Messer® CANGas, 99.999%

Synonym(s):

Oxygen gas

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€182.00

€182.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 L
€182.00

About This Item

Empirical Formula (Hill Notation):
O2
CAS Number:
Molecular Weight:
32.00
EC Number:
MDL number:
UNSPSC Code:
12141904
PubChem Substance ID:
NACRES:
NA.22

€182.00


Please contact Customer Service for Availability

Request a Bulk Order

description

Filling mass: 16g
Filling volume: 10L

Quality Level

Assay

99.999%

form

gas

SMILES string

O=O

InChI

1S/O2/c1-2

InChI key

MYMOFIZGZYHOMD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Oxygen is a colourless, odourless, oxidizing gas that is slightly heavier than air. Examples of applications include special analytical processes (e.g. total organic carbon [TOC] ) and use in the automotive industries. Gas is easy to handle in a small, light pressure can, while the disposable container allows for aluminium recycling when finished.

Recommended products

Z742374: CANgas pressure regulator
Z742379: CANgas plastic hose spout
Z742378: CANgas outer adaptor
Z742377: CANgas inner adaptor
Z742375: CANgas dosing valve

Legal Information

Messer is a registered trademark of Messer Group GmbH

accessory

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Pictograms

Flame over circleGas cylinder

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Gas 1 - Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service