Skip to Content
Merck
All Photos(1)

Documents

769096

Sigma-Aldrich

Air

synthetic, Messer® CANgas, 99.999%

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

vapor density

1 (vs air)

Quality Level

description

Filling mass: 15g
Filling volume: 12L

Assay

99.999%

form

gas

quality

synthetic

bp

−189 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

This synthetic air is a colourless, odourless gas mixture of N2 and O2 with the same weight as air. Applications include gas chromatography with use as a zero gas or oxidizing gas for flame ionization detectors(FID). Gas is easy to handle in a small, light pressure can, while the disposable container allows for aluminium recycling when finished.

Recommended products

Z742374: CANgas pressure regulator
Z742379: CANgas plastic hose spout
Z742378: CANgas outer adaptor
Z742377: CANgas inner adaptor
Z742375: CANgas dosing valve

Legal Information

Messer is a registered trademark of Messer Group GmbH

accessory

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service