Skip to Content
Merck
All Photos(2)

Key Documents

769002

Sigma-Aldrich

Carbon dioxide

Messer® CANgas, 99.995%

Synonym(s):

Carbon dioxide gas

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€195.00

€195.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 L
€195.00

About This Item

Empirical Formula (Hill Notation):
CO2
CAS Number:
Molecular Weight:
44.01
Beilstein:
1900390
EC Number:
MDL number:
UNSPSC Code:
12352119
PubChem Substance ID:
NACRES:
NA.22

€195.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor density

1.52 (vs air)

Quality Level

vapor pressure

56.5 atm ( 20 °C)

description

Filling mass: 24g
Filling volume: 12L

Assay

99.995%

form

gas

mp

−78.5 °C (lit.)

SMILES string

O=C=O

InChI

1S/CO2/c2-1-3

InChI key

CURLTUGMZLYLDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Carbon dioxide is a colourless, odourless, liquefied, and asphyxiating gas that is heavier than air. It is an active gas in CO2 lasers, a component in artificial biological atmospheres, and a solvent for supercritical fluid extraction and chromatography(SFE / SFC). Gas is easy to handle in a small, light pressure can, while the disposable container allows for aluminium recycling when finished.

Recommended products

Z742374: CANgas pressure regulator
Z742379: CANgas plastic hose spout
Z742378: CANgas outer adaptor
Z742377: CANgas inner adaptor
Z742375: CANgas dosing valve

Legal Information

Messer is a registered trademark of Messer Group GmbH

accessory

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

related product

Product No.
Description
Pricing

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service