Skip to Content
Merck
All Photos(2)

Key Documents

65133

Sigma-Aldrich

Methyl 5-methoxy-3-oxovalerate

technical, ≥85% (GC)

Synonym(s):

Methyl 5-methoxy-3-oxopentanoate, Nazarov reagent

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€236.00

€236.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
€236.00

About This Item

Linear Formula:
CH3O(CH2)2COCH2COOCH3
CAS Number:
Molecular Weight:
160.17
Beilstein:
2042099
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€236.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

technical

Assay

≥85% (GC)

form

liquid

refractive index

n20/D 1.44

bp

56-57 °C/0.1 mmHg (lit.)

density

1.10 g/mL at 20 °C (lit.)

functional group

ester
ether
ketone

SMILES string

COCCC(=O)CC(=O)OC

InChI

1S/C7H12O4/c1-10-4-3-6(8)5-7(9)11-2/h3-5H2,1-2H3

InChI key

OPURNNHYTATPKM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Precursor of the unstable methyl 3-oxo-4-pentenoate obtained by acid catalysis (e.g. TosOH) used for annulation reactions. It can be prepared in-situ, review[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K.L. Habermas et al.
Org. React., 45, 1-1 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service