Skip to Content
Merck
All Photos(1)

Documents

165115

Sigma-Aldrich

1,3-Acetonedicarboxylic acid

technical grade

Synonym(s):

3-Oxoglutaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2COCH2COOH
CAS Number:
Molecular Weight:
146.10
Beilstein:
1447081
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Assay

(Remarks on Titration H2SO4 <= 1.0%)

mp

133 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

OC(=O)CC(=O)CC(O)=O

InChI

1S/C5H6O5/c6-3(1-4(7)8)2-5(9)10/h1-2H2,(H,7,8)(H,9,10)

InChI key

OXTNCQMOKLOUAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

392.0 °F - open cup

Flash Point(C)

200 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V N Montesinos et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 15(2), 228-234 (2016-01-13)
Removal of Cr(VI) and citric acid (Cit) by heterogeneous photocatalytic Cr(VI) transformation under UV light over two commercial TiO2 samples (1 g L(-1)), Evonik P25 and Hombikat UV100, was studied at pH 2 and Cr(VI) concentrations between 0.2 and 3
Jian-Ping Huang et al.
Nature communications, 10(1), 4036-4036 (2019-09-08)
The skeleton of tropane alkaloids is derived from ornithine-derived N-methylpyrrolinium and two malonyl-CoA units. The enzymatic mechanism that connects N-methylpyrrolinium and malonyl-CoA units remains unknown. Here, we report the characterization of three pyrrolidine ketide synthases (PYKS), AaPYKS, DsPYKS, and AbPYKS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service