Skip to Content
Merck
All Photos(2)

Key Documents

D138002

Sigma-Aldrich

Dimethyl-1,3-acetonedicarboxylate

96%

Synonym(s):

Dimethyl-3-oxoglutarate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€36.30
100 G
€108.00

€36.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€36.30
100 G
€108.00

About This Item

Linear Formula:
CO(CH2COOCH3)2
CAS Number:
Molecular Weight:
174.15
Beilstein:
1366042
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€36.30


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.444 (lit.)

bp

237-241 °C/760 mmHg (lit.)

density

1.206 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

COC(=O)CC(=O)CC(=O)OC

InChI

1S/C7H10O5/c1-11-6(9)3-5(8)4-7(10)12-2/h3-4H2,1-2H3

InChI key

RNJOKCPFLQMDEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service