Skip to Content
Merck
All Photos(1)

Key Documents

366080

Sigma-Aldrich

2-Amino-3-chloro-5-(trifluoromethyl)pyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4ClF3N2
CAS Number:
Molecular Weight:
196.56
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

86-90 °C (lit.)

functional group

chloro
fluoro

SMILES string

Nc1ncc(cc1Cl)C(F)(F)F

InChI

1S/C6H4ClF3N2/c7-4-1-3(6(8,9)10)2-12-5(4)11/h1-2H,(H2,11,12)

InChI key

WXNPZQIRDCDLJD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterization and luminescence properties of a high quantum efficiency Nd~(3+) organic complex.
Ye Y-X, et al.
Journal of Optoelectronics - Laser / Guang Dian Zi ? Ji Guang, 6, 017-017 (2013)
Luminescent properties of a new Nd3+-doped complex with two different carboxylic acids and pyridine derivative.
Ye Y-X, et al.
Rare Metals, 32(5), 490-495 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service