Skip to Content
Merck
All Photos(1)

Documents

559989

Sigma-Aldrich

2-Chloro-4-(trifluoromethyl)pyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3ClF3N
CAS Number:
Molecular Weight:
181.54
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.4490 (lit.)

bp

146-147 °C (lit.)

density

1.411 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)c1ccnc(Cl)c1

InChI

1S/C6H3ClF3N/c7-5-3-4(1-2-11-5)6(8,9)10/h1-3H

InChI key

GBNPVXZNWBWNEN-UHFFFAOYSA-N

General description

2-Chloro-4-(trifluoromethyl)pyridine can be synthesized from 2-chloro-4-iodopyridine.

Application

2-Chloro-4-(trifluoromethyl)pyridine may be used in the synthesis of:
  • 4,4′-bis( trifluoromethyl)-2,2′-bipyridine
  • 4-(trifluoromethyl)pyridine
  • 1,3-bis(4-(trifluoromethyl)pyridin-2-yl)benzene

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trifluoromethyl-substituted 2, 2'-bipyridine ligands. Synthetic control of excited-state properties of ruthenium (II) tris-chelate complexes.
Furue M, et al.
Inorganic Chemistry, 31(18), 3792-3795 (1992)
Cyclometallated platinum (II) complexes of 1, 3-di (2-pyridyl) benzenes: tuning excimer emission from red to near-infrared for NIR-OLEDs.
Rossi E, et al.
Journal of Materials Chemistry, 21(39), 15501-15510 (2011)
The Direct Metalation and Subsequent Functionalization of Trifluoromethyl-Substituted Pyridines and Quinolines.
Schlosser M and Marull M.
European Journal of Organic Chemistry, 8, 1569-1575 (2003)
Trifluoromethyl-Substituted Pyridines Through Displacement of Iodine by in situ Generated (Trifluoromethyl) copper.
Cottet F and Schlosser M.
European Journal of Organic Chemistry, 2, 327-330 (2002)
Wade C Henke et al.
ChemSusChem, 10(22), 4589-4598 (2017-10-13)
We demonstrate that [Cp*Rh] complexes bearing substituted 2,2'-bipyridyl ligands are effective hydrogen evolution catalysts (Cp*=η

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service