Skip to Content
Merck
All Photos(3)

Key Documents

661120

Sigma-Aldrich

2-Bromo-5-(trifluoromethyl)pyridine

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€29.20
5 G
€83.70

€29.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€29.20
5 G
€83.70

About This Item

Empirical Formula (Hill Notation):
C6H3BrF3N
CAS Number:
Molecular Weight:
225.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€29.20


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

44-48 °C

functional group

bromo
fluoro

SMILES string

FC(F)(F)c1ccc(Br)nc1

InChI

1S/C6H3BrF3N/c7-5-2-1-4(3-11-5)6(8,9)10/h1-3H

InChI key

GSKMWMFOQQBVMI-UHFFFAOYSA-N

Application

Substrate used in a palladium-catalyzed α-arylation of a Refomatsky reagent.[1]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 45, 7395-7395 (2004)
Ben Yiu-Wing Wong et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 22(42), 15095-15106 (2016-09-07)
A series of air-stable spiro-fused ladder-type boron(III) compounds has been designed, synthesized, and the electrochemistry and photophysical behavior have been characterized. By simply varying the substituents on the pyridine ring and extending the π-conjugation of the spiro framework, the emission

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service