Y0002369
Saxagliptin monohydrate
CRS, European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Saxagliptin hydrate, (S)-3-Hydroxyadamantylglycine-L-cis-4,5-methanoprolinenitrile hydrate, BMS 477118, BMS 477118-11, BMS-477118, BMS-477118-11, BMS477118, BMS477118-11
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
saxagliptin
manufacturer/tradename
EDQM
application(s)
pharmaceutical
storage temp.
2-8°C
SMILES string
OC12CC([C@@H](C(N3[C@@H](C[C@]4([C@@]3(C4)[H])[H])C#N)=O)N)(CC(C5)C2)CC5C1.O
InChI
1S/C18H25N3O2.H2O/c19-8-13-2-12-3-14(12)21(13)16(22)15(20)17-4-10-1-11(5-17)7-18(23,6-10)9-17;/h10-15,23H,1-7,9,20H2;1H2/t10?,11?,12-,13+,14+,15-,17?,18?;/m1./s1
InChI key
AFNTWHMDBNQQPX-NHKADLRUSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Biochem/physiol Actions
Packaging
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 2 - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service