Skip to Content
Merck
All Photos(1)

Documents

Y0002361

Supelco

Saxagliptin

for system suitability CRS, European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Saxagliptin hydrate, (S)-3-Hydroxyadamantylglycine-L-cis-4,5-methanoprolinenitrile hydrate, BMS 477118, BMS 477118-11, BMS-477118, BMS-477118-11, BMS477118, BMS477118-11

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H25N3O2 · H2O
CAS Number:
Molecular Weight:
333.43
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

saxagliptin

manufacturer/tradename

EDQM

application(s)

pharmaceutical

storage temp.

2-8°C

SMILES string

OC12CC([C@@H](C(N3[C@@H](C[C@]4([C@@]3(C4)[H])[H])C#N)=O)N)(CC(C5)C2)CC5C1.O

InChI

1S/C18H25N3O2.H2O/c19-8-13-2-12-3-14(12)21(13)16(22)15(20)17-4-10-1-11(5-17)7-18(23,6-10)9-17;/h10-15,23H,1-7,9,20H2;1H2/t10?,11?,12-,13+,14+,15-,17?,18?;/m1./s1

InChI key

AFNTWHMDBNQQPX-NHKADLRUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Saxagliptin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Saxagliptin (BMS-477118) is an orally active, highly potent and long-acting dipeptidyl peptidase IV inhibitor (human DPP-IV Ki = 0.9 nM; plasma DDP-IV inhibition = 87% 0.5-4h post 4 μmol/kg po. in normal rats) with therapeutic efficacy against type 2 diabetes in vivo (plasma glucose clearance = 28/43/61% post 0.3/1.0/3.0 μmol/kg po. in Zucker fa/fa rats).

Packaging

Unit quantity: 1.082mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 2 - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service