Skip to Content
Merck
All Photos(1)

Key Documents

SML3925

Sigma-Aldrich

Liraglutide

≥95% (HPLC)

Synonym(s):

Arg34, Lys26-[Nε(γ-Glu[Nα-hexadecanoyl])]-GLP-1[7-37], H-His-Ala-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Val-Ser-Ser-Tyr-Leu-Glu-Gly-Gln-Ala-Ala-N6-[N-(1-oxohexadecyl)-L-γ-glutamyl]-Lys-α-Glu-Phe-Ile-Ala-Trp-Leu-Val-Arg-Gly-Arg-Gly-OH, HAEGTFTSDVSSYLEGQAA-N6-[N-(1-oxohexadecyl)-L-γ-glutamyl]K-EFIAWLVRGRG, NN2211

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C172H265N43O51
CAS Number:
Molecular Weight:
3751.20
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

(Powder or Lyophilized powder or film)

color

white to off-white

storage temp.

-10 to -25°C

Biochem/physiol Actions

Glucagon-like peptide-1 (GLP-1)-based incretin mimetic with enhanced in vivo stability and efficacy against type 2 diabetes and obesity.
Liraglutide (NN2211) is a glucagon-like peptide-1 (GLP-1)-based incretin mimetic (cAMP stimulation EC50 = 61 nM vs 55 nM with GLP-1(7-37) using human GLP-1 receptor-expressing BHK cells), where GLP-1(7-37) sequence is modified with a Lys34-to-Arg substitution and a γ-Glu-C16 acylation on Lys26 for enhanced in vivo stability (plasma t1/2 in pigs post s.c. = 14 h vs 1.2 h with GLP-1(7-37)) and efficacy against type 2 diabetes and obesity (400 μg/kg/day s.c. in rats; 100-200 μg/kg bid or single 50-200 μg/kg i.v. in rats; 100 μg/kg bid or single 30-1000 μg/kg s.c. in mice; 2 μg/kg i.v. or 3.3 μg/kg/day s.c. in pigs).

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

29953740
Basic and Clinical Pharmacology and Toxicology, 123(6), 704-713 (2018)
Potent derivatives of glucagon-like peptide-1 with pharmacokinetic properties suitable for once daily administration
Journal of Medicinal Chemistry, 43(9), 1664-1669 (2000)
NN2211: a long-acting glucagon-like peptide-1 derivative with anti-diabetic effects in glucose-intolerant pigs
European Journal of Pharmacology, 451(2), 217-225 (2002)
GLP-1 Analogue Liraglutide Enhances SP-A Expression in LPS-Induced Acute Lung Injury through the TTF-1 Signaling Pathway
Mediators of Inflammation, 2018, 3601454-3601454 (2018)
Liraglutide improves cognitive impairment via the AMPK and PI3K/Akt signaling pathways in type 2 diabetic rats
Molecular Medicine Reports, 18(2), 2449-2457 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service