Skip to Content
Merck
All Photos(1)

Key Documents

GE17-5474-02

MabSelect SuRe LX

Cytiva 17-5474-02, pack of 200 mL

Synonym(s):

Protein purification resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56
Pricing and availability is not currently available.

ligand

alkali-stabilized protein A-derived (E. coli)

packaging

pack of 200 mL

manufacturer/tradename

Cytiva 17-5474-02

storage condition

(20% Ehtanol)

matrix

highly cross-linked agarose

avg. part. size

85 μm (d₅₀v is the median particle size of the cumulative volume distribution)

cleaning in place

2-13.7

working range

3-12

capacity

~60 mg binding capacity

suitability

suitable for bioprocess medium

storage temp.

2-8°C

Related Categories

General description

MabSelect SuRe LX is a protein A affinity medium with very high dynamic binding capacity at extended residence times and is developed for high titer antibody processes. Alkali tolerance, high capacity and low ligand leakage in combination with the rigid base matric makes MabSelect SuRe LX highly suitable for purification of monoclonal anitbodies for clinical applications.

MabSelect SuRe LX is a member of the MabSelect family of affinity chromatography media for the capture of monoclonal antibodies (MAbs) at process scale. MabSelect SuRe LX shares features and benefits of MabSelect and MabSelect SuRe such as a rigid, high-flow agarose matrix that ensures excellent pressure/flow properties, low non-specific binding that leads to low impurity levels in the eluate pool, and an oriented ligand coupling for optimal binding capacity. It also features the same alkali-tolerant rProtein A ligand as MabSelect SuRe. This ligand provides greater stability than conventional protein A-based media in the alkaline conditions used in cleaning-in-place (CIP) and sanitization protocols. The enhanced alkali stability of MabSelect SuRe improves process economy and product quality; cleaning can be performed with cost-effective reagents such as sodium hydroxide eliminating the need for expensive and hazardous cleaning agents such as Gua-HCl. Process economy is also improved by the increased dynamic binding capacity of MabSelect SuRe LX.

Just as for MabSelect SuRe, the novel ligand construct of MabSelect SuRe LX has been proven to have an increased stability towards proteases. Therefore, the level of leakage of the MabSelect SuRe LX ligand during elution is low.

MabSelect SuRe has been demonstrated to give generic elution conditions for different monoclonal antibodies which can be applied also to MabSelect SURe LX since it utilizes the same ligand. This is an advantage when designing generic purification platform processes. The feature is also correlated to the construction of the MabSelect SuRe ligand.

MabSelect SuRe LX is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, MabSelect SuRe meets industrial demands with security of supply and comprehensive technical and regulatory support.
human IgG/ml medium. Determined at 10 % breakthrough by frontal analysis at a mobile phase velocity of 100 cm/h in a column with a bed height of 20 cm, i.e. residence time is 6.0 min.
pH below 3 is sometimes required to elute strongly bound IgG species. However, protein ligands may hydrolyze at very low pH.

Features and Benefits

  • Novel, alkali-tolerant rProtein A ligand withstands rigorous CIP and sanitization procedures with 0.1 to 0.5 M NaOH.
  • Reduces the risk of product contamination and carryover
  • Improves performance and reduces overall costs
  • High dynamic binding capacity (DBC) for high titer cultures reduces process time and amount of medium used
  • Generic elution profile for different monoclonal antibodies enables platform approach to purification
  • High-flow agarose matrix allows the processing of large feed volumes

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Free MabSelect SuRe Ligand
"MabSelect SuRe Ligand Restricted License" and "Cys-rProtein A Ligand Restricted License" are protected by the following patents and equivalent patents and patent applications in other countries: US 6,399,750 and EP 1123389. A free, non-transferable limited license to use this product for internal analytical purposes only accompanies the purchase of the product from a Cytiva company and its licensed distributors. Any other use will require a separate license from a Cytiva company.
MabSelect is a trademark of Cytiva
MabSelect SuRe is a trademark of Cytiva
SuRe is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Capto adhere from Cytiva is a multimodal strong anion exchanger for BioProcess applications.

Description of viral clearance using Capto adhere from Cytiva.

This page describes the difference between bind/elute and flowthrough modes in multimodal chromatography using Cytiva products.

This page describes the maintenance of media and storage conditions for multimodal chromatography using Cytiva products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service