Skip to Content
Merck
All Photos(1)

Key Documents

GE17-5438-02

MabSelect SuRe

Cytiva 17-5438-02, pack of 200 mL

Synonym(s):

protein A affinity medium, affinity chromatography media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56
Pricing and availability is not currently available.

ligand

alkali-stabilized protein A-derived (E. coli)

packaging

pack of 200 mL

manufacturer/tradename

Cytiva 17-5438-02

storage condition

(20% Ehtanol)

matrix

highly cross-linked agarose

avg. part. size

85 μm (d₅₀v is the median particle size of the cumulative volume distribution)

cleaning in place

2-13.7

working range

3-12

capacity

≥30 mg binding capacity (humanlgG/ml at 2.4 min residence time)

suitability

suitable for bioprocess medium

storage temp.

2-8°C

Related Categories

General description

Determined at 10% breakthrough by frontal analysis at a mobile phase velocity of 500 cm/h in a column with a bed height of 20 cm.
MabSelect SuRe is a protein A affinity medium for capturing MAbs at process scale. MabSelect SuRe has an alkali-tolerant rProtein A ligand that allows the use of rigorous and cost-effective CIP and sanitization protocols based on 0.1 to 0.5 M NaOH. Further, the ligand is protease stable leading to lower ligand leakage.

MabSelect SuRe is a member of the MabSelect family of affinity chromatography media for the capture of monoclonal antibodies (MAbs) at process scale. MabSelect SuRe shares features and benefits of MabSelect such as a rigid, high-flow agarose matrix that ensures excellent pressure/flow properties, low non-specific binding that leads to low impurity levels in the eluate pool, and an oriented ligand coupling for optimal binding capacity. In addition, MabSelect SuRe features an alkali-tolerant rProtein A ligand. This ligand provides greater stability than conventional protein A-based media in the alkaline conditions used in cleaning-in-place (CIP) and sanitization protocols. The enhanced alkali stability of MabSelect SuRe improves process economy and product quality; cleaning can be performed with cost-effective reagents such as sodium hydroxide eliminating the need for expensive and hazardous cleaning agents such as Gua-HCl.

The novel ligand construct of MabSelect SuRe has been proven to have an increased stability towards proteases. Therefore, the level of leakage of the MabSelect SuRe ligand during elution is low. MabSelect SuRe has been demonstrated to give generic elution conditions for different monoclonal antibodies, which is an advantage when designing generic purification platform processes. This feature is also correlated to the construction of the MabSelect SuRe ligand.

MabSelect SuRe is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, MabSelect SuRe meets industrial demands with security of supply and comprehensive technical and regulatory support.
pH below 3 is sometimes required to elute strongly bound IgG species. However, protein ligands may hydrolyze at very low pH.

Features and Benefits

  • Novel, alkali-tolerant rProtein A ligand withstands rigorous CIP and sanitization procedures with 0.1 to 0.5 M NaOH.
  • Reduces the risk of product contamination and carryover
  • Improves product quality and reduces overall costs
  • Novel ligand design gives enhanced protease resistance resulting in lower ligand leakage
  • Generic elution profile for different monoclonal antibodies enables platform approach to purification
  • Used in the commercial manufacture of monoclonal antibodies
  • High-flow agarose matrix allows the processing of large feed volumes

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Free MabSelect SuRe Ligand
"MabSelect SuRe Ligand Restricted License" and "Cys-rProtein A Ligand Restricted License" are protected by the following patents and equivalent patents and patent applications in other countries: US 6,399,750 and EP 1123389. A free, non-transferable limited license to use this product for internal analytical purposes only accompanies the purchase of the product from a Cytiva company and its licensed distributors. Any other use will require a separate license from a Cytiva company.
MabSelect is a trademark of Cytiva
SuRe is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Description of viral clearance using Capto adhere from Cytiva.

Capto adhere from Cytiva is a multimodal strong anion exchanger for BioProcess applications.

This page describes the difference between bind/elute and flowthrough modes in multimodal chromatography using Cytiva products.

This page describes the maintenance of media and storage conditions for multimodal chromatography using Cytiva products.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service