Skip to Content
Merck
All Photos(1)

Key Documents

GE17-5468-02

Capto SP ImpRes

Cytiva 17-5468-02, pack of 100 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47101511
NACRES:
NA.56
Pricing and availability is not currently available.

ligand

sulfonate group

description

Ion Exchanger Type (value)

packaging

pack of 100 mL

manufacturer/tradename

Cytiva 17-5468-02

matrix

high-flow agarose

avg. part. size

36-44 μm (d50v)

cleaning in place

3-14

working range

4-12

capacity

>70 mg binding capacity(> 70 mg lysozyme, > 95 mg BSA)

suitability

suitable for bioprocess medium

General description

Capto SP ImpRes is a strong cation exchange BioProcess medium, designed to meet the demands of modern large-scale manufacturers for fast, efficient, and cost-effective intermediate and polishing protein purification.

Application

Capto SP ImpRes is a strong cation exchanger for the high-throughput intermediate purification and polishing steps of a wide range of biomolecules. This chromatography media is part of an expanded high-resolution platform based on the high-fLow Agarose Capto product line.

By combining the high-fLow characteristics of Capto media with a small particle size, Capto SP ImpRes delivers excellent pressure-fLow properties with impressive resolution. The ability to run at higher flow rates and higher bed heights also increases flexibility in process design.

Capto SP ImpRes is a BioProcess chromatography medium developed for large-scale purification of biopharmaceutical Proteins. Based on the well-established Capto base matrix and with traditional ionic group ligands, it provides many opportunities for improved productivity and straightforward process development.
Capto SP ImpRes is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development.
As member of the BioProcess media range, Capto SP ImpRes meets industrial demands with security of supply and comprehensive technical and regulatory support.

Features and Benefits

  • High-resolution intermediate purification and polishing media based on the well-established Capto platform with traditional ligands.
  • Flexible process design due to large operational window of flow rates and bed heights.
  • High throughput purifications easy to optimize and scale-up.
  • Higher manufacturing productivity enables improved process economy.
  • BioProcess media developed and supported for large-scale chromatography.
  • Prepacked HiTrap® columns for easy screening and small-scale purification.
  • Security of supply and comprehensive regulatory support

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol + 0.2 M Sodium Acetate)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Capto is a trademark of Cytiva
HiTrap is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page describes the maintenance of media and storage conditions for multimodal chromatography using Cytiva products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service