Key Documents
GBL664113
Grace Bio-Labs FastWells reagent barriers
1- 20 mm diam. x 1.0 mm depth
Synonym(s):
flexible silicone slide gasket, reagent chamber
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
Pricing and availability is not currently available.
feature
wells 1
packaging
pack of 50 ea
manufacturer/tradename
Grace Bio-Labs 664113
external size
25 mm × 25 mm
Compare Similar Items
View Full Comparison
Show Differences
1 of 4
This Item
GBL664114GBL204908GBL664112
packaging
pack of 50 ea
packaging
pack of 50 ea
packaging
pack of 10 ea
packaging
pack of 50 ea
feature
wells 1
feature
wells 1
feature
wells 8
feature
wells 1
external size
25 mm × 25 mm
external size
25.5 mm × 44 mm
external size
-
external size
-
General description
FastWells reagent barriers are sticky, flexible silicone gaskets that form hydrophobic barriers around specimens without messy adhesives or special slides. Gaskets may be stacked to increase depth and volume.
FastWells provide rapid isolation of cells and tissues on slides or coverslips for antibody incubations. The barriers remain sealed during agitated washing steps, preventing reagents from spilling when shaken or rocked. FastWells peel off quickly and cleanly, leaving no residue to interfere with specimen cover-slipping. They can be washed, autoclaved and reused. The barriers can also be sealed to form incubation chambers using flexible HybriSlip covers.
Legal Information
FastWells is a trademark of Grace Bio-Labs, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)