Key Documents
GBL204908
Grace Bio-Labs FlexWell removable incubation chambers
L × W × thickness 6.5 mm × 6.5 mm × 3.2 mm, clear silicone, adhesive one side, wells, 8
Synonym(s):
flexible silicone slide gasket, silicone multiwell gasket
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
Pricing and availability is not currently available.
material
clear silicone
sterility
non-sterile
feature
wells 8
packaging
pack of 10 ea
manufacturer/tradename
Grace Bio-Labs 204908
L × W × thickness
6.5 mm × 6.5 mm × 3.2 mm
General description
FlexWell incubation chambers provide a convenient way to conduct binding reactions on protein microarrays using nitrocellulose coated ONCYTE® Film slides.
  • Silicone FlexWell forms wells on slides using a clean release adhesive to isolate up to 16 specimens per slide
  • Slides and FlexWell fit into a reusable slide tray organizing wells into the footprint of a SMS compliant microtiter plate
  • Gaskets may be sealed for mixing and to prevent evaporation
Legal Information
FlexWell is a trademark of Grace Bio-Labs, Inc.
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Hallo, Ich bin auf der Suche nach einem Produkt, mit dem ich Kammern auf meinen Objektträgern bilden kann. Sind das bodenlose Kammern, so dass ich sie auf einen Objektträger kleben kann, um eine Brunnenkammer auf meinen Objektträgern zu bilden?
    1 answer
    1. This product is manufactured by Grace Bio Labs and their catalog number is 204908. For assistance with your stated application on this third-party product, please contact the manufacturer directly at 1-800-813-7339.
      Helpful?