Skip to Content
Merck
All Photos(1)

Key Documents

Y0000452

Morphine sulfate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Morphine sulfate salt pentahydrate, Morphine hemi[sulfate pentahydrate]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H40N2O10S · 5H2O
CAS Number:
Molecular Weight:
758.83
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

morphine

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O.O.O.O.O.OS(O)(=O)=O.CN1CC[C@]23[C@H]4Oc5c(O)ccc(C[C@@H]1[C@@H]2C=C[C@@H]4O)c35.CN6CC[C@]78[C@H]9Oc%10c(O)ccc(C[C@@H]6[C@@H]7C=C[C@@H]9O)c8%10

InChI

1S/2C17H19NO3.H2O4S.5H2O/c2*1-18-7-6-17-10-3-5-13(20)16(17)21-15-12(19)4-2-9(14(15)17)8-11(10)18;1-5(2,3)4;;;;;/h2*2-5,10-11,13,16,19-20H,6-8H2,1H3;(H2,1,2,3,4);5*1H2/t2*10-,11+,13-,16-,17-;;;;;;/m00....../s1

InChI key

GRVOTVYEFDAHCL-RTSZDRIGSA-N

Gene Information

human ... OPRM1(4988)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Morphine sulfate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hélène Mottaz et al.
Environmental pollution (Barking, Essex : 1987), 221, 105-115 (2016-12-25)
Opioid drugs, such as morphine (MO), detected in aquatic environments worldwide, may harm fish due to their semi-persistence and ability to potently interact with molecular targets conserved across vertebrates. Here, we established a waterborne bacterial lipopolysaccharide (LPS) challenge assay with
Nian Gong et al.
Anesthesiology, 120(4), 962-975 (2013-08-10)
D-Amino acid oxidase (DAAO) is a flavin adenine dinucleotide-dependent peroxisomal flavoenzyme which is almost exclusively expressed within astrocytes in the spinal cord. DAAO catalyzes oxidation of D-amino acids to hydrogen peroxide, which is a stable and less active reactive oxygen
Serotonin, morphine, and neuropathic pain: not a simple story.
Kelly Byrne et al.
Anesthesiology, 121(2), 217-218 (2014-06-03)
Ehab Farag et al.
Anesthesiology, 121(2), 239-248 (2014-04-25)
Ultrasound guidance for continuous femoral perineural catheters may be supplemented by electrical stimulation through a needle or through a stimulating catheter. The authors tested the primary hypothesis that ultrasound guidance alone is noninferior on both postoperative pain scores and opioid
L Zoric et al.
British journal of anaesthesia, 112(4), 722-728 (2014-01-17)
The infiltration of local anaesthetic (LA), ketorolac, and epinephrine has been suggested to be effective for analgesia after total hip arthroplasty (THA). The part of action of each component of the mixture remains unclear. We investigated the contribution of infiltration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service