
Morphine sulfate

Synonym(s):
Morphine sulfate salt pentahydrate, Morphine hemi[sulfate pentahydrate]
Empirical Formula (Hill Notation):
(C17H19NO3)2 · H2SO4 · 5H2O
CAS Number:
Molecular Weight:
758.83
Products
Formula Weight
Gene Alias
Manufacturer
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2