Skip to Content
Merck
All Photos(1)

Key Documents

PHR8861

Morphine sulfate

certified reference material, pharmaceutical secondary standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
(C17H19NO3)2 · H2SO4 · 5H2O
CAS Number:
Molecular Weight:
758.83
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

API family

morphine

CofA

current certificate can be downloaded

packaging

pkg of 1 g

application(s)

pharmaceutical

storage temp.

-10 to -25°C

SMILES string

[S](=O)(=O)(O)O.N6([C@H]7[C@H]8[C@@]9([C@@H](Oc%10c9c(ccc%10O)C7)[C@H](C=C8)O)CC6)C.N1([C@H]2[C@H]3[C@@]4([C@@H](Oc5c4c(ccc5O)C2)[C@H](C=C3)O)CC1)C.O.O.O.O.O

InChI key

GRVOTVYEFDAHCL-RTSZDRIGSA-N

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Signal Word

Warning

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service