Skip to Content
Merck
All Photos(1)

Key Documents

Z413097

Sigma-Aldrich

Aldrich® micro distilling splitter head

Jacketed, Overall H 260 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104812
NACRES:
NB.43
Pricing and availability is not currently available.

Overall H

260 mm

immersion depth

50 mm

Looking for similar products? Visit Product Comparison Guide

General description

The column and receiver joints are 14/20 with a 10/18 thermometer joint, two 2 mm glass stopcocks, PTFE 0-3 mm metering valve. Removable condenser cold-finger for easy cleaning.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service