Key Documents
Z174513
Sigma-Aldrich
Aldrich® micro short-path distillation apparatus
Standard head
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
Pricing and availability is not currently available.
Related Categories
General description
Threaded 14/20 joints. Thermometers are included.
Replacement parts:
Z174521 - Standard head
Z247774 - Jacketed head
Z174548 - Cow receiver
Z174556 - Recovery flask, 5 mL
Z174564 - Distillation flask, 10 mL
Z174572 - Gas-inlet tube
Z561487 - Thermometer, 25 mm immr. L, -10 to 250 °C
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)