Key Documents
Z127779
Sigma-Aldrich
Aldrich® fraction cutter head
joint: ST/NS 24/40
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
Pricing and availability is not currently available.
Joint
joint: ST/NS24/40
General description
50 mL barrel, with 4 mm bore glass stopcocks.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)