Skip to Content
Merck
All Photos(1)

Key Documents

Y0001628

Tribenoside impurity D

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Benzyl ether, Dibenzyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2)2O
CAS Number:
Molecular Weight:
198.26
Beilstein:
1911156
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

tribenoside

manufacturer/tradename

EDQM

refractive index

n20/D 1.561 (lit.)

bp

298 °C (lit.)

mp

1.5-3.5 °C (lit.)

density

1.043 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C(OCc1ccccc1)c2ccccc2

InChI

1S/C14H14O/c1-3-7-13(8-4-1)11-15-12-14-9-5-2-6-10-14/h1-10H,11-12H2

InChI key

MHDVGSVTJDSBDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tribenoside impurity D EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

278.6 °F - Pensky-Martens closed cup

Flash Point(C)

137 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin Matt et al.
Angewandte Chemie (International ed. in English), 53(46), 12446-12450 (2014-09-10)
Organizing magnetic nanoparticles into long-range and dynamic assemblies would not only provide new insights into physical phenomena but also open opportunities for a wide spectrum of applications. In particular, a major challenge consists of the development of nanoparticle-based materials for
Dan Heo et al.
Nanotechnology, 25(27), 275102-275102 (2014-06-25)
In this study, we developed the maleimidyl magnetic nanoplatform, which enables functional targeting of a biomarker-specific moiety for molecular imaging via MRI. The maleimide group of the maleimidyl magnetic nanoplatform is conjugated with a thiol group without additional crosslinkers and
Sasmita Mohapatra et al.
Dalton transactions (Cambridge, England : 2003), 43(42), 15841-15850 (2014-09-17)
The development of theranostic mesoporous hollow silica nanospheres having therapeutic and diagnostic functions has been achieved. The exterior surface of the hollow mesoporous silica nanosphere was selectively acid functionalized and utilised to conjugate the anticancer drug cisplatin, the marker molecule
Jin Han et al.
ACS nano, 9(3), 2805-2819 (2015-02-18)
Electrophysiological phenotype development and paracrine action of mesenchymal stem cells (MSCs) are the critical factors that determine the therapeutic efficacy of MSCs for myocardial infarction (MI). In such respect, coculture of MSCs with cardiac cells has windowed a platform for
Lijiao Ao et al.
Nanoscale, 6(18), 10710-10716 (2014-08-07)
This paper devotes a novel micellar structure for cancer theranostics by incorporating magnetic and therapeutic functionalities into a natural sourced targeting polymer vehicle. Heparin-folic acid micelles taking advantage of both excellent loading capability and cancer targeting ability have been employed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service