Merck
Maleimidyl magnetic nanoplatform for facile molecular MRI.
Nanotechnology (2014-06-25)
Dan Heo, Eugene Lee, Minhee Ku, Seungyeon Hwang, Bongjune Kim, Yeonji Park, Young Han Lee, Yong-Min Huh, Seungjoo Haam, Jae-Ho Cheong, Jaemoon Yang, Jin-Suck Suh
ABSTRACT
In this study, we developed the maleimidyl magnetic nanoplatform, which enables functional targeting of a biomarker-specific moiety for molecular imaging via MRI. The maleimide group of the maleimidyl magnetic nanoplatform is conjugated with a thiol group without additional crosslinkers and side products. A physicochemical analysis was conducted to verify the effectiveness of the maleimidyl magnetic nanoplatform, and the existence of the maleimidyl group was investigated using the platform. To prepare biomarker-specific MRI probes, a thiolated aptamer and peptide were immobilized onto the maleimidyl group of the maleimidyl magnetic nanoplatform. The fabricated MRI probes were applied to four cancer cell lines: HT1080, MCF7, MKN45, and HEK293T. To investigate the potential of the molecular MRI probe, the target-biomarker specificity was confirmed without serious cytotoxicity, and in vivo MRI analysis using a xenograft mouse model was demonstrated. We believe these results will be useful for fabricating molecular MRI probes for the diagnosis of cancer.
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Dodecylamine, 98%
Sigma-Aldrich
Triethylamine, purum, ≥99% (GC)
Sigma-Aldrich
Dodecylamine, puriss., ≥99.5% (GC)
Sigma-Aldrich
Dodecylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Supelco
Triethylamine, analytical standard
Valaciclovir impurity G, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Triethylamine, ≥99.5%
USP
Valacyclovir Related Compound G, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Benzyl ether, ≥98%, FCC, FG
Sigma-Aldrich
Benzyl ether, 98%
Sigma-Aldrich
Dibenzyl ether, purum, ≥98.0% (GC)
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
4-(Dimethylamino)pyridine, purum, ≥98.0% (NT)
Sigma-Aldrich
Manganese(II) acetylacetonate
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
DCC, 99%
Sigma-Aldrich
DCC, puriss., ≥99.0% (GC)
Sigma-Aldrich
DCC, 1.0 M in methylene chloride
Sigma-Aldrich
N-Maleoyl-β-alanine, 97%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, analytical standard
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer