Merck
  • Soft magnets from the self-organization of magnetic nanoparticles in twisted liquid crystals.
Soft magnets from the self-organization of magnetic nanoparticles in twisted liquid crystals.
Angewandte Chemie (International ed. in English) (2014-09-10)
Benjamin Matt, Kirsten M Pondman, Sarah J Asshoff, Bennie Ten Haken, Benoit Fleury, Nathalie Katsonis
ABSTRACT
Organizing magnetic nanoparticles into long-range and dynamic assemblies would not only provide new insights into physical phenomena but also open opportunities for a wide spectrum of applications. In particular, a major challenge consists of the development of nanoparticle-based materials for which the remnant magnetization and coercive field can be controlled at room temperature. Our approach consists of promoting the self-organization of magnetic nanoparticles in liquid crystals (LCs). Using liquid crystals as organizing templates allows us to envision the design of tunable self-assemblies of magnetic nanoparticles, because liquid crystals are known to reorganize under a variety of external stimuli. Herein, we show that twisted liquid crystals can be used as efficient anisotropic templates for superparamagnetic nanoparticles and demonstrate the formation of hybrid soft magnets at room temperature.
Product Description
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Oleylamine, technical grade, 70%
Sigma-Aldrich
Benzyl ether, 98%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Benzyl ether, ≥98%, FCC, FG
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Dibenzyl ether, purum, ≥98.0% (GC)
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, natural, FCC
Supelco
Dichloromethane, analytical standard
Supelco
Oleic acid, Selectophore, ≥99%
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
SAFC
Oleylamine, ≥98% (primary amine)
Tribenoside impurity D, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer