Skip to Content
Merck
All Photos(1)

Key Documents

790625P

Avanti

DC-Chol/DOPE Blend

Avanti Research - A Croda Brand 790625P, powder

Synonym(s):

DC-Chol/DOPE Blend, DC-Cholesterol/Dioleoyl Phosphatidylethanolamine (30:70, w/w)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 25 mg (790625P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 790625P

lipid type

cationic lipids
transfection

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

DC-Cholesterol/Dioleoyl Phosphatidylethanolamine (DOPE) lipid complex comprises the cationic lipid DC-Cholesterol and the neutral helper lipid DOPE.

Application

DC-Chol/DOPE Blend has been used:
  • in the preparation of liposomes for ferritin heavy chain 1 (FTH1) siRNA (small interference RNA) transfection studies
  • to compare its cytotoxicity tests with ginger derived nanoparticles in colon-26 cells
  • as a standard control in MTT (3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyl tetrazolium bromide) assay to evaluate biocompatibility of ginger derived lipid vesicles

Biochem/physiol Actions

DC-Cholesterol/Dioleoyl Phosphatidylethanolamine (30:70, w/w) helps in transfection of therapeutic small interference RNA (siRNA). DC-Chol/DOPE liposomes are cationic, unilamellar and spherical in nature. It aids in improved condensation of DNA for transfection studies.

Packaging

5 mL Clear Glass Sealed Ampule (790625P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingzhen Zhang et al.
Nanomedicine (London, England), 12(16), 1927-1943 (2017-07-01)
To develop novel siRNA delivery system overcoming the limitations of synthetic nanoparticles, such as potential side effects, nonspecificity and economic production for ulcerative colitis therapy. Nanoparticles composed of edible ginger-derived lipid, termed ginger-derived lipid vehicles (GDLVs) were generated from ginger
Meike-Kristin Abraham et al.
Nanotheranostics, 1(2), 154-165 (2017-10-27)
Rationale: Genetic therapy using modified mRNA for specific therapeutic protein expression for disease treatment and vaccination represents a new field of therapeutic and diagnostic medicine. Non-viral vectors transfection using biocompatible nanoliposomes enables safe and efficient delivery of therapeutic mRNA. Objective:
Mingzhen Zhang et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 24(10), 1783-1796 (2016-08-06)
The use of nanotechnology for drug delivery has shown great promise for improving cancer treatment. However, potential toxicity, hazardous environmental effects, issues with large-scale production, and potential excessive costs are challenges that confront their further clinical applications. Here, we describe
Alberto Rodríguez-Pulido et al.
The journal of physical chemistry. B, 112(39), 12555-12565 (2008-08-30)
A 1:1 mixture of the cationic lipid 3beta-[ N-( N', N'-dimethylaminoethane)-carbamoyl]cholesterol hydrochloride (DC-Chol) and the zwitterionic lipid, 1,2-dioleoyl- sn-glycero-3-phosphoetanolamine (DOPE), has been used to compact calf-thymus DNA (CT-DNA) in aqueous buffered solution at 298.15 K. The formation process of this
Vagisha Ravi et al.
PloS one, 14(9), e0221952-e0221952 (2019-09-07)
Elevated expression of the iron regulatory protein, ferritin heavy chain 1 (FTH1), is increasingly being associated with high tumor grade and poor survival outcomes in glioblastoma. Glioma initiating cells (GICs), a small population of stem-like cells implicated in therapeutic resistance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service