Skip to Content
Merck
All Photos(1)

Key Documents

790625C

Avanti

DC-Chol/DOPE Blend

Avanti Research - A Croda Brand 790625C

Synonym(s):

DC-Chol/DOPE Blend, DC-Cholesterol/Dioleoyl Phosphatidylethanolamine (30:70, w/w)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 2.5 mL (790625C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 790625C

concentration

10 mg/mL (790625C-25mg)

lipid type

cationic lipids

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

DC-Chol/DOPE blend is a cationic lipid blend.
DC-Chol/DOPE is a blend of a cationic lipid : 3b-[N-(N′, N′ dimethyl-aminoethane)carbomyl] (DC-Chol ) and a neutral lipid : dioleoyl phosphatidylethanolamine (DOPE).

Biochem/physiol Actions

The DC-Chol/DOPE blend is widely useful in liposome preparation as it influences the process of encapsulation. 1:2 or 3:2 is a preferred molar ratio for DC-Chol/DOPE liposomes.

Packaging

5 mL Clear Glass Sealed Ampule (790625C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aspartoacylase gene transfer to the mammalian central nervous system with therapeutic implications for Canavan disease
Leone P, et al.
Annals of Neurology, 48(1), 27-38 (2000)
Shuoye Yang et al.
International journal of pharmaceutics, 434(1-2), 155-160 (2012-05-31)
Three most commonly used preparative methods, dry-film, reverse phase evaporation and ethanol injection were employed to prepare cationic liposomes composed of DC-Chol and DOPE, respectively. The resulting samples were contrasted through morphology observation, particle size and zeta potential analysis. Sephadex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service