Skip to Content
Merck
All Photos(1)

Key Documents

890840P

Avanti

14:0 DAP

Avanti Research - A Croda Brand 890840P, powder

Synonym(s):

1,2-dimyristoyl-3-dimethylammonium-propane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H65NO4
CAS Number:
Molecular Weight:
539.87
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 25 mg (890840P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 890840P

lipid type

cationic lipids
transfection

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(CN(C)C)(OC(CCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCC)=O

Application

1,2-dimyristoyl-3-dimethylammonium-propane (14:0 DAP) might be used:
  • to prepare liposomal membrane and study its interaction between surfactants
  • to study its effect on the ordering of two-dimensional dispersions of membrane-coated particles
  • to study its interaction with mastoparan (MP)

Biochem/physiol Actions

1,2-dimyristoyl-3-dimethylammonium-propane (14:0 DAP) is a cationic lipid and serves as a pH-sensitive transfection reagent.

Packaging

5 mL Clear Glass Sealed Ampule (890840P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Nomura et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(5), 2340-2345 (2001-02-28)
Dynamic behaviors of liposomes caused by interactions between liposomal membranes and surfactant were studied by direct real-time observation by using high-intensity dark-field microscopy. Solubilization of liposomes by surfactants is thought to be a catastrophic event akin to the explosion of
Specific ion interaction dominates over hydrophobic matching effects in peptide-lipid bilayer interactions: the case of short peptide
Wei F, et al.
The Journal of Physical Chemistry C, 117(49), 26190-26196 (2013)
Esther W Gomez et al.
Soft matter, 5(9), 1931-1936 (2009-05-07)
Two-dimensional dispersions of colloidal particles with a range of surface chemistries and electrostatic potentials are characterized under a series of solution ionic strengths. A combination of optical imaging techniques are employed to monitor both the colloid structure and the electrostatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service