Skip to Content
Merck
All Photos(1)

Key Documents

09-0610

Sigma-Aldrich

Ethanolamine hydrochloride

SAJ special grade, ≥98.0%

Synonym(s):

2-Aminoethanol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2CH2OH · HCl
CAS Number:
Molecular Weight:
97.54
Beilstein:
3542892
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:

grade

SAJ special grade

Assay

≥98.0%

form

solid

availability

available only in Japan

mp

82-84 °C (lit.)

SMILES string

NCCO.[H]Cl

InChI

1S/C2H7NO.ClH/c3-1-2-4;/h4H,1-3H2;1H

InChI key

PMUNIMVZCACZBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hua Xu et al.
Bio-medical materials and engineering, 26 Suppl 1, S2091-S2100 (2015-09-26)
In this paper, we report the development of a protein microarray-based biosensor for the detection of the hepatitis B virus (HBV) serological markers using surface plasmon resonance (SPR Printing buffer, protein immobilization time and concentration of the capture protein were
Dae-Ho Jang et al.
Colloids and surfaces. B, Biointerfaces, 123, 887-891 (2014-12-04)
A sensitive method for selectively detecting insulin-like growth factor-binding protein 7 (IGFBP7) over a wide range of concentrations based on the surface plasmon resonance (SPR) biosensing techniques is described. IGFBP7 has been shown to regulate cell proliferation, cell adhesion, cellular
P Rockenfeller et al.
Cell death and differentiation, 22(3), 499-508 (2015-01-13)
Autophagy is a cellular recycling program that retards ageing by efficiently eliminating damaged and potentially harmful organelles and intracellular protein aggregates. Here, we show that the abundance of phosphatidylethanolamine (PE) positively regulates autophagy. Reduction of intracellular PE levels by knocking
Hiroki Kinouchi et al.
Analytical biochemistry, 492, 43-48 (2015-09-20)
We developed a surface plasmon resonance (SPR) assay to estimate the competitive inhibition by pharmaceuticals for thyroxine (T4) binding to thyroid hormone transport proteins, transthyretin (TTR) and thyroxine binding globulin (TBG). In this SPR assay, the competitive inhibition of pharmaceuticals
Se Shi et al.
Biosensors & bioelectronics, 74, 454-460 (2015-07-15)
A sensitive and stable electroless-plated gold film for the preparation of an optical fiber surface plasmon resonance (SPR) sensor is presented in this work, together with a facile antibody immobilization method. Gold nanoparticles were uniformly adsorbed onto the surface of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service