Merck
  • Multiplex biomarker analysis biosensor for detection of hepatitis B virus.
Multiplex biomarker analysis biosensor for detection of hepatitis B virus.
Bio-medical materials and engineering (2015-09-26)
Hua Xu, Dayong Gu, Jian'an He, Lei Shi, Jingyu Yao, Chunxiao Liu, Chunzhong Zhao, Yunqing Xu, Shengyang Jiang, Jun Long
ABSTRACT
In this paper, we report the development of a protein microarray-based biosensor for the detection of the hepatitis B virus (HBV) serological markers using surface plasmon resonance (SPR Printing buffer, protein immobilization time and concentration of the capture protein were optimized systematically to determine the best performance of the biosensor. Under optimal conditions, five hepatitis B markers in 20 μL human serum can be simultaneously detected within 30 minutes, whereas other methods such as ELISA and PCR can detect only one marker within four hours. This platform has been validated by analysis of 35 patients known to have hepatitis B, with 85% agreement between the test platform and analysis by commercial enzyme-linked immunosorbent assay (ELISA) kits. The results demonstrate that the protein microarray with SPR displayed a sensitivity of 0.1 ng mL(-1) for HBsAg. In addition to high sensitivity, it also shows excellent specificity, reproducibility and stability. This integrated protein microarray technique combined with SPR is a promising candidate for hepatitis B diagnosis with high-throughput.
Sigma-Aldrich
Ethanolamine hydrochloride, ≥99.0%
Sigma-Aldrich
Ethanolamine hydrochloride, SAJ special grade, ≥98.0%
Sigma-Aldrich
1,2-Dichloroethane, SAJ first grade, ≥99.0%
Sigma-Aldrich
1,2-Dichloroethane, JIS special grade, ≥99.5%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Ethanolamine hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%