Skip to Content
AW
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ethanolamine hydrochloride
Synonym(s):
2-Aminoethanol hydrochloride
Linear Formula:
H
2
NCH
2
CH
2
OH · HCl
CAS Number:
2002-24-6
Molecular Weight:
97.54
EC Number:
217-900-6
Products
Product Category
buffers (2)
building blocks (1)
Brand
Sigma-Aldrich (3)
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
pK Value
Product Line
Purity
Research Area
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Anhydrous (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
E6133
Ethanolamine hydrochloride,
≥99.0%
Expand
View Pricing
RDD028
Ethanolamine hydrochloride,
anhydrous, free-flowing, Redi-Dri
™
, ≥99.0%
Expand
View Pricing
09-0610
Ethanolamine hydrochloride,
SAJ special grade, ≥98.0%
Expand
View Pricing