Skip to Content
Merck
All Photos(3)

Key Documents

V800087

Sigma-Aldrich

Bromocresol Green

Powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H14Br4O5S
CAS Number:
Molecular Weight:
698.01
Beilstein:
372527
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Product Name

Bromocresol Green,

product line

Vetec

form

powder

visual transition interval

3.8-5.4, yellow to blue

mp

225 °C (dec.) (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cc1c(Br)c(O)c(Br)cc1C2(OS(=O)(=O)c3ccccc23)c4cc(Br)c(O)c(Br)c4C

InChI

1S/C21H14Br4O5S/c1-9-12(7-14(22)19(26)17(9)24)21(13-8-15(23)20(27)18(25)10(13)2)11-5-3-4-6-16(11)31(28,29)30-21/h3-8,26-27H,1-2H3

InChI key

FRPHFZCDPYBUAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janeta Starosvetsky et al.
Journal of microbiological methods, 92(2), 178-182 (2012-11-28)
High concentrations of H(2)S in groundwater are commonly removed using Biological Trickling Filter (BTF) that contains high numbers of biofilm immobilized sulfur oxidizing bacteria (mainly Thiobacillus thiooxidans). BTF performance requires continuous monitoring of these bacteria at several sampling points. The
Yu-Chang Chang et al.
Sensors (Basel, Switzerland), 11(4), 4060-4072 (2011-12-14)
A meso-structured Al-MCM-41 material was impregnated with bromocresol green (BG) dye and then incorporated into a UV-Vis DRA spectroscopic instrument for the online detection of ammonia gas. The absorption response of the Al-MCM-41/BG ammonia sensing material was very sensitive at
Kai Yu et al.
Enzyme and microbial technology, 49(3), 272-276 (2011-11-25)
A pH-sensitive colorimetric assay has been established to quantitatively measure glutamate decarboxylase (GAD) activity in bacterial cell extracts using a microplate format. GAD catalyzes the irreversible α-decarboxylation of L-glutamate to γ-aminobutyrate. The assay is based on the color change of
Kohji Hirata et al.
Research communications in molecular pathology and pharmacology, 117-118, 115-124 (2008-04-23)
Administration of antitumor drugs such as 5-fluorouracil and methotrexate to rats causes intestinal injury. The intestinal injury was reported to be prevented by coadministration of prostaglandin E1 analogues to rats. IEC-6 cell is a useful in vitro model to study
Carolyn Cray et al.
Journal of avian medicine and surgery, 25(1), 14-22 (2011-06-11)
The purpose of this study was to examine the validity of albumin determinations in penguin plasma by the bromcresol green (BCG) method and the gold standard of protein electrophoresis (EPH). Plasma from 96 clinically normal and abnormal penguins (Spheniscus species)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service