Skip to Content
Merck
All Photos(2)

Key Documents

318728

Sigma-Aldrich

Bromocresol Green/Methyl Red, mixed indicator solution

in methanol

Synonym(s):

Tetrabromocresolsulfonphthalein

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

form

liquid

Quality Level

composition

Bromocresol Green, 0.1%
Methyl Red, 0.02%

concentration

in methanol

density

0.839 g/mL at 25 °C

λmax

434 nm

ε (extinction coefficient)

≥10000 at 282-288 nm in methanol
≥20000 at 509-519 nm in methanol
≥7000 at 329-335 nm in methanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CN(C)c1ccc(cc1)\N=N\c2ccccc2C(O)=O.Cc3c(Br)c(O)c(Br)cc3C4(OS(=O)(=O)c5ccccc45)c6cc(Br)c(O)c(Br)c6C

InChI

1S/C21H14Br4O5S.C15H15N3O2/c1-9-12(7-14(22)19(26)17(9)24)21(13-8-15(23)20(27)18(25)10(13)2)11-5-3-4-6-16(11)31(28,29)30-21;1-18(2)12-9-7-11(8-10-12)16-17-14-6-4-3-5-13(14)15(19)20/h3-8,26-27H,1-2H3;3-10H,1-2H3,(H,19,20)/b;17-16+

InChI key

MFUUASJGLBWWPF-ZYSYNQLUSA-N

General description

Mixed indicator solutions of bromocresol green (pk 4.9) and methyl red (pk 5) give a red color in acidic conditions and green color in basic conditions with a sharp transition through grey color at pH 5.1 as the two colors are complementary.[1]

Application

Suitable for titration of morpholine

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S M Khopkar
Basic Concepts Of Analytical Chemistry (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service