Skip to Content
Merck
All Photos(3)

Documents

250198

Sigma-Aldrich

Methyl Red

ACS reagent, crystalline

Synonym(s):

2-(4-Dimethylaminophenylazo)benzoic acid, 4-Dimethylaminoazobenzene-2′-carboxylic acid, Acid Red 2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4N=NC6H4CO2H
CAS Number:
Molecular Weight:
269.30
Colour Index Number:
13020
Beilstein:
750102
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

ACS reagent

Quality Level

form

crystalline

technique(s)

titration: suitable

visual transition interval

4.2-6.2, pink to yellow

mp

179-182 °C (lit.)

solubility

ethanol: 1 mg/mL

λmax

410 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CN(C)c1ccc(cc1)\N=N\c2ccccc2C(O)=O

InChI

1S/C15H15N3O2/c1-18(2)12-9-7-11(8-10-12)16-17-14-6-4-3-5-13(14)15(19)20/h3-10H,1-2H3,(H,19,20)/b17-16+

InChI key

CEQFOVLGLXCDCX-WUKNDPDISA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Methyl Red has been used as pH indicator in a glucose biosensor, which is used for salivary analysis.

Biochem/physiol Actions

Methyl Red is a maroon red crystal azo dye. Methyl Red is a pH indicator and changes color at a pH of 5.5. Methyl Red is widely used in saliva sampling method. In addition, it is also employed in carbohydrate and lactic acid detection. Methyl Red has been effectively used for intrageneric differentiation of Enterobacteriaceae.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A paper strip based non-invasive glucose biosensor for salivary analysis.
Soni A
Biosensors And Bioelectronics, 67, 763-768 (2015)
R R colwell and R grigorova
Methods in Microbiology, 19 (1987)
R. W. Sabnis
Handbook of Acid-Base Indicators (2007)
An improved dithizone method for the determination of small quantities of zinc in blood and tissue samples.
Vallee BL and Gibson JG 2nd
The Journal of Biological Chemistry, 176, 435-443 (1948)
Oana Alexandru et al.
Journal of neuro-oncology, 102(1), 9-18 (2010-07-17)
A major focus of brain cancer research today is to translate understanding of glioma biology into advances in treatment, by exploring the potential of target therapy. Here we investigated the ability of three compounds belonging to the chemical class of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service