Skip to Content
Merck
All Photos(1)

Documents

V800011

Sigma-Aldrich

1-Pentanesulfonic acid sodium salt

suitable for HPLC, ≥99%

Synonym(s):

Sodium pentanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4SO3Na
CAS Number:
Molecular Weight:
174.19
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

product line

Vetec

Assay

≥99%

technique(s)

HPLC: suitable

mp

>300 °C (lit.)

SMILES string

[Na+].CCCCCS([O-])(=O)=O

InChI

1S/C5H12O3S.Na/c1-2-3-4-5-9(6,7)8;/h2-5H2,1H3,(H,6,7,8);/q;+1/p-1

InChI key

ROBLTDOHDSGGDT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joe Z Sostaric
Ultrasonics sonochemistry, 15(6), 1043-1048 (2008-05-13)
Sonolysis of aqueous solutions of n-alkyl anionic surfactants results in the formation of secondary carbon-centered radicals (-*CH-). The yield of -*CH- depends on the bulk surfactant concentration up to a maximum attainable radical yield (the 'plateau yield') where an increasing
Limei Yang et al.
The journal of physical chemistry. B, 111(6), 1361-1367 (2007-01-26)
Sonolysis of argon-saturated aqueous solutions of the nonvolatile surfactants sodium dodecyl sulfate (SDS) and sodium 1-pentanesulfonate (SPSo) was investigated at three ultrasonic frequencies under both continuous wave (CW) and pulsed ultrasound. Secondary carbon-centered radicals were detected by spin trapping using
B R Thomas et al.
Journal of pharmaceutical and biomedical analysis, 12(1), 85-90 (1994-01-01)
The five active drug substances and two of the excipients present in Frenadol, a cold medication, were separated. The active drug components dextromethorphan HBr monohydrate, ascorbic acid, caffeine, paracetamol and chlorpheniramine maleate were quantitatively assayed by a mixed ion pair
Hsin-Hua Yeh et al.
Electrophoresis, 26(4-5), 947-953 (2005-01-26)
A simple and selective micellar electrokinetic chromatography (MEKC) with UV detection is described for simultaneous determination of amikacin, tobramycin, and kanamycin A, performed in Tris buffer (180 mM; pH 9.1) with 300 mM sodium pentanesulfonate (SPS) as an anionic surfactant.
Rahmat Sadeghi et al.
Journal of colloid and interface science, 346(1), 107-117 (2010-03-24)
Thermodynamic properties for aqueous solutions containing sodium pentane-1-sulfonate (C(5)SO(3)Na) in the absence and presence of poly(ethyleneglycol) (PEG) or poly(vinylpyrrolidone) (PVP) determined as a function of surfactant concentration from the density, sound velocity, viscosity, conductivity and vapor-liquid equilibria data, are reported

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service