Skip to Content
Merck
All Photos(4)

Key Documents

O8380

Sigma-Aldrich

1-Octanesulfonic acid sodium salt

≥98%

Synonym(s):

Octane-1-sulfonic acid sodium salt, Sodium 1-octanesulfonate, Sodium octane-1-sulfonate, Sodium octanesulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$75.60
25 G
$227.00
100 G
$743.00

$75.60


Available to ship on04 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
$75.60
25 G
$227.00
100 G
$743.00

About This Item

Linear Formula:
C8H17O3SNa
CAS Number:
Molecular Weight:
216.27
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.21

$75.60


Available to ship on04 April 2025Details


Request a Bulk Order

biological source

synthetic (oragnic)

Quality Level

description

anionic

Assay

≥98%

form

powder

mol wt

216.27 g/mol

technique(s)

HPLC: suitable

impurities

<2.0% water (Karl Fischer)

mp

300  °C (572 °F)

solubility

9.8 g/L at 20 °C

cation traces

Na: 10.1-11.1 % (w/v) (Anhydrous)

SMILES string

[Na+].CCCCCCCCS([O-])(=O)=O

InChI

1S/C8H18O3S.Na/c1-2-3-4-5-6-7-8-12(9,10)11;/h2-8H2,1H3,(H,9,10,11);/q;+1/p-1

InChI key

HRQDCDQDOPSGBR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

1-Octanesulfonic acid sodium is an anionic detergent and a surfactant with excellent coupling properties. This versatile molecule finds application as a buffer component, in cell biology, analytical chemistry, and biochemical research.

Application

1-Octanesulfonic acid sodium salt has been used:
  • as a component of 9 parts buffer to determine the concentration of L-Dopa in extracts of seed flour
  • in mobile phase solution to measure dopamine in rats using high-performance liquid chromatography (HPLC) analysis
  • in neurochemistry for the preparation of mobile phase buffer for HPLC analysis.

Ion-pairing reagent for HPLC analysis of peptides and proteins.

Features and Benefits

  • Highly versatile surfactant for your cell biology and biochemical research
  • Suitable for HPLC applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

comparable product

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I T James et al.
Journal of chromatography, 525(1), 43-57 (1990-01-26)
Following a detailed study, a rapid and sensitive assay for the naturally fluorescent collagen cross-links pyridinoline and deoxypyridinoline has been developed using ion-pair reversed-phase high-performance liquid chromatography in the presence of 1-octanesulphonic acid (OSA). Pyridinoline and deoxypyridinoline were separated on
P G Jaison et al.
Journal of chromatography. A, 1216(9), 1383-1389 (2009-01-27)
Studies were carried out for the separation of uranium (U) and thorium (Th) on reversed-phase (RP) C18 columns using mandelic acid as an eluent. Retention of thorium-mandelate on the unmodified stationary phase was found to be greater than that of
Tejwant Singh et al.
Physical chemistry chemical physics : PCCP, 12(37), 11728-11735 (2010-08-10)
We have observed dual transitions in various physical properties while investigating the surfactant-like behavior of the ionic liquid (IL) 1-butyl-3-methylimidazolium octylsulfate [C(4)mim][C(8)SO(4)] in aqueous media. Interestingly, in conductivity measurements, it was found that the second transition, which is comparatively weak
Yingjie Ma et al.
Chemical communications (Cambridge, England), 47(45), 12340-12342 (2011-10-21)
By the introduction of trimethylammonium groups at both upper and lower rims, a cationic water-soluble pillar[5]arene was prepared, which forms a stable 1 : 1 host-guest complex with sodium 1-octanesulfonate in water.
J G Foulks et al.
Canadian journal of physiology and pharmacology, 63(10), 1327-1334 (1985-10-01)
The potency of various types of alkyl amphipathic (cationic, anionic, and neutral) as well as tertiary amine local anesthetics in impairing the excitability of frog skeletal muscle was markedly enhanced by an increase in temperature from 20 to 30 degrees

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service