Skip to Content
Merck
All Photos(1)

Key Documents

1.18304

Supelco

Pentane-1-sulfonic acid sodium salt

for ion pair chromatography LiChropur

Synonym(s):

Pentane-1-sulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$1,040.00
100 G
$3,130.00

$1,040.00


Available to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
$1,040.00
100 G
$3,130.00

About This Item

Empirical Formula (Hill Notation):
C5H11NaO3S
CAS Number:
Molecular Weight:
174.19
MDL number:
UNSPSC Code:
12352107
EC Index Number:
245-208-4
NACRES:
NA.21

$1,040.00


Available to ship on02 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥99% (acidimetric)

technique(s)

ion pair chromatography: suitable

loss

≤2% loss on drying, 1200°C; 4h; vacuum

transmittance

200 nm, ≥70%
220 nm, ≥90%
250 nm, ≥98%
(0.005 M; 1 cm in H2O)

pH

5.5-7.5 (20 °C, 100 g/L in H2O)

bulk density

390 kg/m3

suitability

passes test for identity (IR)

storage temp.

15-25°C

SMILES string

[Na+].[S](=O)(=O)([O-])CCCCC

InChI

1S/C5H12O3S.Na/c1-2-3-4-5-9(6,7)8;/h2-5H2,1H3,(H,6,7,8);/q;+1/p-1

InChI key

ROBLTDOHDSGGDT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium pentanesulfonate is commonly used as an anionic ion-pairing reagent in ion-pair chromatography. It is widely used in the analysis of basic compounds by chromatographic analysis.[1][2]

Application

Pentane-1-sulfonic acid sodium salt can be used as anion-pair reagent in the:

  • Method for determining gemcitabine and its metabolite, dFdU.[1]
  • Analysis of alkaloid standards.[3]

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, structure elucidation, determination of the lipophilicity and identification of antitumour activities in vitro of novel 3-(2-furanyl)-8-aryl-7, 8-dihydroimidazo [2, 1-c][1, 2, 4] triazin-4 (6H)-ones with a low cytotoxicity towards normal human skin fibroblast cells
Sztanke K, et al.
Bioorganic & Medicinal Chemistry, 19(17), 5103-5116 (2011)
Effect of chromatographic conditions on the separation and system efficiency for HPLC of selected alkaloids on different stationary phases
Petruczynik A
Journal of AOAC (Association of Official Analytical Chemists) International, 94(1), 77 -789 (2011)
Development and validation of an ion pair HPLC method for gemcitabine and 2?, 2?-difluoro-2?-deoxyuridine determination
Losa R, et al.
Analytica Chimica Acta, 528(2), 255-260 (2005)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service