Skip to Content
Merck
All Photos(1)

Key Documents

Z290874

Frit for Supelco® Filter

stainless steel, pore size 0.5 μm, pkg of 5 ea

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
$90.50

$90.50


Available to ship on23 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 EA
$90.50

About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

$90.50


Available to ship on23 April 2025Details


Request a Bulk Order

material

stainless steel

packaging

pkg of 5 ea

pore size

0.5 μm

Looking for similar products? Visit Product Comparison Guide

Application

This product may be used to protect HPLC columns containing 5 μm or larger particles against particulate matter that gets accumulated during the chromatographic operations.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

replacement part

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service