Skip to Content
Merck
All Photos(1)

Key Documents

59038

Column Frits

diam. 1/4 in., porosity 0.5 μm

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$82.30

$82.30


Available to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 EA
$82.30

About This Item

UNSPSC Code:
41115700

$82.30


Available to ship on14 April 2025Details


Request a Bulk Order

description

for Conventional Column Hardware (compression style)

packaging

pkg of 10 ea

diam.

1/4 in.

thickness

0.39 in.

pore size

0.5 μm porosity

compatibility

for use with compression style column hardware

Looking for similar products? Visit Product Comparison Guide

General description

Choose frits with 0.5 μm pores to retain packings prepared from 3 μm particles, frits with 2 μm pores to retain larger particles.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. What is the frit material for part number 59038?'

    1 answer
    1. The frits are made of stainless steel.

      Helpful?

  2. What is the material for the column frits? Are they a silica based frit or are there any metal or other additives (exact composition is not needed just base material)? Thank you very much for you answer and time!

    1 answer
    1. The frits are made of stainless steel.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service