Skip to Content
Merck
All Photos(1)

Key Documents

502790

Frits for Precolumn MicroFilter

PEEK, pore size 0.5 μm

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$188.00

$188.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 EA
$188.00

About This Item

UNSPSC Code:
41115700

$188.00


Check Cart for Availability

Request a Bulk Order

material

PEEK

packaging

pkg of 10 ea

manufacturer/tradename

Upchurch Scientific, Inc A-735X

frit W × D × H

0.045 in. × 0.031 in. × 0.192 in.

pore size

0.5 μm

compatibility

for use with Precolumn Microfilters Assembly

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Hola, necesito comprar un PEEK inline filter para un sistema AKTA, pero dudo de si este producto se corresponde con el que necesito. Se trata del disco que va dentro del filtro (cilindro color crudo) o la pieza cilíndrica en sí misma? Muchas gracias!

    1 answer
    1. This is a filter that mounts in a filter assembly. This product is not compatible with AKTA systems. Please contact your regional Technical Service group for a filter recommendation for the specific application.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service