Skip to Content
Merck
All Photos(1)

Key Documents

Z227137

Nucleosil® Guard Column

phase C18 (octadecyl), 5 μm particle size, L × I.D. 1 cm × 4.6 mm

Sign Into View Organizational & Contract Pricing

Select a Size

4 EA
$639.00

$639.00


Available to ship on22 April 2025Details



Select a Size

Change View
4 EA
$639.00

About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

$639.00


Available to ship on22 April 2025Details


Agency

suitable for USP L1

Quality Level

product line

Nucleosil®

feature

endcapped

packaging

pkg of 4 ea

manufacturer/tradename

Nucleosil®

extent of labeling

15% Carbon loading

parameter

400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

1 cm × 4.6 mm

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

100 Å

compatibility

for use with 3.0 to 4.6 mm column

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

For all guard cartridges listed here use holder 54987

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Nucleosil is a registered trademark of Macherey-Nagel GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service