Skip to Content
Merck
All Photos(1)

Key Documents

56130AST

Supelco

apHera C18 (5 μm) HPLC Columns

L × I.D. 1 cm × 4.6 mm, HPLC Guard Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$989.00

$989.00


Check Cart for Availability


Select a Size

Change View
1 EA
$989.00

About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

$989.00


Check Cart for Availability

Product Name

apHera C18 HPLC Guard Column, 5 μm particle size, L × I.D. 1 cm × 4.6 mm

material

stainless steel column

Quality Level

Agency

suitable for USP L67

product line

apHera

packaging

pkg of 1 ea

manufacturer/tradename

apHera

parameter

147 bar pressure (2133 psi)
4-60 °C temperature

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

1 cm × 4.6 mm

matrix

polymeric

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

300 Å

operating pH

1-14

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

apHera is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service