Skip to Content
Merck
All Photos(1)

Key Documents

50164-U

Nucleosil® C18 HPLC Column

pore size 100 Å, 5 μm particle size, L × I.D. 12.5 cm × 4.6 mm

Synonym(s):

Octadecyl Silica Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,130.00

$1,130.00


Available to ship on14 April 2025Details



Select a Size

Change View
1 EA
$1,130.00

About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

$1,130.00


Available to ship on14 April 2025Details


Agency

suitable for USP L1

Quality Level

product line

Nucleosil®

feature

endcapped

manufacturer/tradename

Nucleosil®

extent of labeling

15% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

12.5 cm × 4.6 mm

surface area

350 m2/g

matrix

octadecyl

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

100 Å

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Related Categories

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Nucleosil is a registered trademark of Macherey-Nagel GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service