Key Documents
SU860065
Supelco
Headspace vial, beveled top, rounded bottom
volume 5 mL, beveled finish clear glass, O.D. × H × I.D. 22 mm × 38.2 mm × 12.5 mm, pkg of 100 ea
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
material
beveled finish clear glass
round bottom
feature
closure type crimp top vial
packaging
pkg of 100 ea
O.D. × H × I.D.
22 mm × 38.2 mm × 12.5 mm
volume
5 mL
fitting
for crimp seal
Choose from one of the most recent versions:
Certificates of Analysis (COA)