Skip to Content
Merck
All Photos(1)

Key Documents

30226-U

Supelco

Supel-Inert Multi-Layer Foil

maximum volume 1 L, Screw Cap Valve (SCV), pkg of 10 bags

Synonym(s):

Supel-Inert Foil 1L SCV Gas Sampling Bag with Thermogreen® LB-2 Septa

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$224.00

$224.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 EA
$224.00

About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

$224.00


Available to ship on22 April 2025Details


Request a Bulk Order

Agency

NIOSH 6603
OSHA ID-172,ID-210

description

Chemically Inert with Light and Moisture Protection
septum replaceable; Thermogreen® LB-2 septa

product line

Supel

packaging

pkg of 10 bags

technique(s)

whole air sampling: suitable

maximum volume

1 L

size × W × L

× 7 in. × 9 in.

suitability

suitable for (Biogas and Landfill Gas (LFG))
suitable for (Calibration Mixes and Gas Transfer)
suitable for (Low molecular weight compounds such as methane, CO, CO2, and permanent gases)
suitable for

application(s)

air monitoring
environmental
industrial hygiene

Looking for similar products? Visit Product Comparison Guide

General description

Multi-Layer Foil: (1) 0.002" Polyethylene (2) 0.0003" Aluminum Foil (3) Polyethylene (4) Aluminum (metalized) (5) 60 Gauge Nylon
Sulfur Compounds: carbon disulfide, hydrogen sulfide, t-butyl mercaptan, ethyl mercaptan, isopropyl mercaptan, n-propyl mercaptan
SupelInert multi-layer foil gas sampling bags with Thermogreen LB-2 septa are ideal for sampling low molecular weight compounds such as carbon monoxide (CO), carbon dioxide (CO2) and permanent gases. They are also suitable for sampling sulfur compounds such as hydrogen sulfide (H2S), carbon disulfide (CS2) and mercaptans.

Key Features and Benefits:

  • Multi-layer foil composed of two outer layers of aluminum film providing a great barrier to gases permeating through walls of the bag
  • Proven low-bleed Thermogreen LB-2 septa installed in the valve fitting
  • Chemically inert with moisture and light protection
  • Not recommended for low ppm VOCs due to background levels (we recommend the SupelInert PVDF Tedlar alternative film for VOCs)
  • Two valve choices available: Screw Cap Valve (SCV) and Push/Pull Lock Valve (PLV)
  • 4 sizes: 1 L, 2 L, 5 L, and 10 L

Legal Information

Supel is a trademark of Sigma-Aldrich Co. LLC
Thermogreen is a registered trademark of Merck KGaA, Darmstadt, Germany

air sampling device

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How permeable are they? We are looking at storing hydrogen for 2/3 days in these bags. Is this viable ?

    1 answer
    1. The bags are not permeable and are suitable for storing gases for 2-3 days. However, caution must be observed so as not to over-fill the bags. All gas sampling bags (Tedlar and Supel-Inert) should only be filled to 80% of the volume to prevent overpressure. Filling the bag with pressures greater than 1 psi (~0.07 bar) could cause the bag to burst or leak the sample.

      Helpful?

  2. Do you sell the septa of these bags separately as well? The manual says septa can be replaced, and the bag reused even after the septum has too many punctures and doesn't seal properly anymore. If you do, I'd like to order septa only.

    1 answer
    1. Yes, the septa replacement product number is 29458-U. Please see the link below to access the replacement:
      https://www.sigmaaldrich.com/product/supelco/29458u

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service