Skip to Content
Merck
All Photos(1)

Key Documents

BP444

4-Methyl-5-(2-hydroxyethyl)thiazole

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

4-Methyl-5-thiazoleethanol, 5-(2-Hydroxyethyl)-4-methylthiazole, Sulfurol

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$384.00

$384.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$384.00

About This Item

Empirical Formula (Hill Notation):
C6H9NOS
CAS Number:
Molecular Weight:
143.21
Beilstein:
114249
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$384.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

clomethiazole, thiamine

form

liquid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

refractive index

n20/D 1.550 (lit.)

bp

135 °C/7 mmHg (lit.)

density

1.196 g/mL at 25 °C (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

Cc1ncsc1CCO

InChI

1S/C6H9NOS/c1-5-6(2-3-8)9-4-7-5/h4,8H,2-3H2,1H3

InChI key

BKAWJIRCKVUVED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

4-Methyl-5-(2-hydroxyethyl)thiazole BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Packaging

Unit quantity: 25 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service